missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-m-xylene, 95%, Thermo Scientific™

Product Code. 10784943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10784943 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10784943 Supplier Thermo Scientific Acros Supplier No. 412500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-28-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00013706
InChI Key BUNKQJAMHYKQIM-UHFFFAOYSA-N
Synonym 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr
PubChem CID 77885
IUPAC Name 1-iodo-2,4-dimethylbenzene
SMILES CC1=CC=C(I)C(C)=C1

Specifications

CAS Min % 94.0
Color Yellow
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5970 to 1.5990 (20°C, 589nm)
Assay Percent Range 95%
Formula Weight 232.06
Percent Purity ≥94%
Chemical Name or Material 4-Iodo-m-xylene, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.