missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-D-phenylalanine, 95%, Thermo Scientific™

Product Code. 15482659
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62561-75-5
Molecular Formula C9H10INO2
Molecular Weight (g/mol) 291.09
MDL Number MFCD00063067
InChI Key PZNQZSRPDOEBMS-MRVPVSSYSA-N
Synonym 4-iodo-d-phenylalanine, h-d-phe 4-i-oh, d-4-iodophenylalanine, d-phe 4-i-oh, p-iodo-d-phenylalanine, d-4-iodophe, r-2-amino-3-4-iodophenyl propanoic acid, d-4-iodo phenylalanine, 2r-2-amino-3-4-iodophenyl propanoic acid, d-4-iodophenyl alanine
PubChem CID 2733279
SMILES [NH3+][C@H](CC1=CC=C(I)C=C1)C([O-])=O

Specifications

Melting Point 251°C (decomposition)
Color Yellow
IUPAC Name (2R)-2-amino-3-(4-iodophenyl)propanoic acid
Formula Weight 291.09
Percent Purity 95%
Quantity 25 g
Sensitivity Light sensitive
Physical Form Powder
Chemical Name or Material 4-Iodo-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'