missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-5-methyl-isoxazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 4-iodo-5-methyl-1,2-oxazole |
Formula Weight | 208.98 |
Percent Purity | 97% |
Chemical Name or Material | 4-Iodo-5-methyl-isoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'