missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-5-methyl-isoxazole, 97%, Thermo Scientific™

Product Code. 10187942
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7064-38-2
Molecular Formula C28H36N4O6S2
Molecular Weight (g/mol) 588.74
MDL Number MFCD06411541
InChI Key GDARZPKSXSXRDC-JCMHNJIXNA-N
Synonym 4-iodo-5-methylisoxazole, 4-iodo-5-methyl-isoxazole, isoxazole,4-iodo-5-methyl, isoxazole, 4-iodo-5-methyl
PubChem CID 2795461
SMILES CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCCCCC(O)=O)C2=O)=C1N1CCCC(C1)C(=O)OCC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 4-iodo-5-methyl-1,2-oxazole
Formula Weight 208.98
Percent Purity 97%
Chemical Name or Material 4-Iodo-5-methyl-isoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'