missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-(trifluoromethyl)-1-H-pyrazole, 97%, Thermo Scientific™

Product Code. 11976231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11976231 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11976231 Supplier Thermo Scientific Acros Supplier No. 444820010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 866638-72-4
Melting Point 139.0°C to 141.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H2F3IN2
Quantity 1 g
Synonym 4-iodo-3-trifluoromethyl-1h-pyrazole, 4-iodo-5-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-iodo-3-trifluoromethyl, acmc-209qaw, 4-iodo-3-trifluoromethyl pyrazole, 3-trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3-trifluoromethyl-2h-pyrazole
InChI Key XRUHSOXIZMYOHX-UHFFFAOYSA-N
SMILES C1=NNC(=C1I)C(F)(F)F
IUPAC Name 4-iodo-5-(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol) 261.97
PubChem CID 16069792
Formula Weight 261.97
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodo-3-(trifluoromethyl)-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.