missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-methyl-5-phenylisoxazole, 97%, Thermo Scientific™

Product Code. 10089111
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089111 10 g Amber Glass Bottle 10g
10478182 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089111 Supplier Thermo Scientific Maybridge Supplier No. CC20311EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16114-53-7
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD03086126
InChI Key RLZGZJOMCXKYIW-UHFFFAOYSA-N
Synonym 4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
PubChem CID 2776520
IUPAC Name 4-iodo-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1I)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-methyl-5-phenylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.