missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-(trifluoromethyl)benzonitrile, 97%

Product Code. p-7037050
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15481928

Brand: Thermo Scientific Alfa Aesar H55609.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 101066-87-9 | C8H3F3IN | 297.019 g/mol

4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101066-87-9
Molecular Formula C8H3F3IN
Molecular Weight (g/mol) 297.019
MDL Number MFCD08458091
InChI Key RBRTXKYFMXVIER-UHFFFAOYSA-N
Synonym 4-iodo-2-trifluoromethyl benzonitrile, 2-cyano-5-iodobenzotrifluoride, benzonitrile, 4-iodo-2-trifluoromethyl, 2-trifluoromethyl-4-iodobenzonitrile, 4-cyano-3-trifluoromethyl-1-iodobenzene, 4-cyano-3-trifluoromethyl iodobenzene, 4-iodo-2-trifluoromethyl benzenecarbonitrile, pubchem4810, acmc-1bqw7, ksc494m4b
PubChem CID 16218337
IUPAC Name 4-iodo-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1I)C(F)(F)F)C#N

Specifications

Melting Point 50°C to 55°C
Flash Point >110°C (230°F)
Quantity 250 mg
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 297.02
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.