missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-methylaniline, 98+%, Thermo Scientific™

Product Code. 11397655
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13194-68-8
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00025299
InChI Key BGKLFAQCHHCZRZ-UHFFFAOYSA-N
Synonym 2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
PubChem CID 83221
IUPAC Name 4-iodo-2-methylaniline
SMILES CC1=C(C=CC(=C1)I)N

Specifications

Melting Point 84°C to 88°C
Quantity 25 g
UN Number UN2811
Beilstein 2353618
Sensitivity Light sensitive
Formula Weight 233.05
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-2-methylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'