missing translation for 'onlineSavingsMsg'
Learn More

4-iodo-2-methyl-1H-imidazole, Thermo Scientific™

Product Code. 10406862
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10406862 10 g Amber Glass Bottle 10g
10513232 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406862 Supplier Thermo Scientific Maybridge Supplier No. MO00831EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 73746-45-9
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD02684335
InChI Key SEDSLMWYUQACGR-UHFFFAOYSA-N
PubChem CID 2794623
IUPAC Name 5-iodo-2-methyl-1H-imidazole
SMILES CC1=NC=C(I)N1

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Synonym 4-iodo-2-methyl-1h-imidazole, 2-methyl-4 5-iodo-1 h-imidazole, 4-iodo-2-methylimidazole, 2-methyl-4-iodo-1h-imidazole, 4-iodo-2-methyl-3h-imidazole, 1h-imidazole, 4-iodo-2-methyl, 2-methyl-4-jodimidazol, ksc638k7d, 1h-imidazole,4-iodo-2-methyl, 1h-imidazole,5-iodo-2-methyl
Formula Weight 208.0016
Chemical Name or Material 4-iodo-2-methyl-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.