missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2,6-dimethylphenol, 97%, Thermo Scientific™

Product Code. 10101322
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10101322 50 g 50g
10533242 1 g 1g
10262362 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10101322 Supplier Thermo Scientific Maybridge Supplier No. RDP00223EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10570-67-9
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.06
MDL Number MFCD00094415
InChI Key HUUNIMCCAGNBDF-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-iodophenol, 4-iodo-2,6-dimethyl-phenol, 4-iodo-2,6-xylenol, acmc-2098hg, phenol,4-iodo-2,6-dimethyl, huunimccagnbdf-uhfffaoysa, phenol, 4-iodo-2,6-dimethyl
PubChem CID 555318
IUPAC Name 4-iodo-2,6-dimethylphenol
SMILES CC1=CC(I)=CC(C)=C1O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
Formula Weight 248.06
Percent Purity 97%
Chemical Name or Material 4-Iodo-2, 6-dimethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.