missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-iodo-1,2-dimethylbenzene, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Formula Weight | 232.06 |
Percent Purity | 97% |
Density | 1.6300g/mL |
Quantity | 25 g |
Chemical Name or Material | 4-Iodo-o-xylene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'