missing translation for 'onlineSavingsMsg'
Learn More

4-Indolylboronic acid, 95%, Thermo Scientific™

Product Code. 15254358 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15254358

Brand: Thermo Scientific Acros 454500010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 220465-43-0
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.97
InChI Key USJUQEVUEBCLLR-UHFFFAOYSA-N
Synonym indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid
PubChem CID 2763173
IUPAC Name 1H-indol-4-ylboronic acid
SMILES B(C1=C2C=CNC2=CC=C1)(O)O

Specifications

Melting Point 184.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 160.97
Percent Purity 95%
Chemical Name or Material 4-Indolylboronic acid
compliance-icons
Product Identifier
  • 4-Indolylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.