missing translation for 'onlineSavingsMsg'
Learn More

4-IBP, MedChemExpress

Product Code. 16452808
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452808 10 mg 10mg
16495547 5 mg 5mg
16439486 50 mg 50mg
16404246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452808 Supplier Medchem Express Supplier No. HY100155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-IBP is a selective σ1 agonist with a high level of affinity for the σ1 receptor (Ki = 1.7 nM) and a moderate affinity for the σ2 receptor (Ki = 25.2 nM) .

4-IBP is a selective σ1 agonist with a high level of affinity for the σ1 receptor (Ki = 1.7 nM) and a moderate affinity for the σ2 receptor (Ki = 25.2 nM) .

Specifications

Chemical Name or Material 4-IBP
CAS 155798-08-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21IN2O
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (237.93 mM; Need ultrasonic)
SMILES O=C(NC1CCN(CC2=CC=CC=C2)CC1)C3=CC=C(I)C=C3
Molecular Weight (g/mol) 420.29
Formula Weight 420.29
Percent Purity 98.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.