missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

Product Code. 10755191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10755191 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10755191 Supplier Thermo Scientific Acros Supplier No. 121910010
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 345909-35-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 275.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H7NO3
MDL Number MFCD00149476
Quantity 1 g
Beilstein 22, 230
Merck Index 14, 5327
Synonym 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
Solubility Information Solubility in water: 0.9g/L water (100°C). Other solubilities: soluble in hot alcohol, insoluble in ether, very soluble in alkali
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
SMILES OC(=O)C1=NC2=CC=CC=C2C(O)=C1
IUPAC Name 4-oxo-1H-quinoline-2-carboxylic acid;hydrate
Molecular Weight (g/mol) 189.17
PubChem CID 18530299
Formula Weight 189.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxyquinoline-2-carboxylic acid, hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.