missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinoline-2-carboxylic acid hydrate, 98%

Product Code. 11424334
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424334 5 g 5g
11414334 1 g 1g
11434334 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424334 Supplier Thermo Scientific Alfa Aesar Supplier No. A12602.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol

Broad spectrum excitatory amino acid antagonist4-Hydroxyquinoline-2-carboxylic acid hydrate is used as an intermediate in synthetic chemistry. It acts as an antiexcitotoxic and anticonvulsant. It finds application in certain neurobiological disorders therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot ethanol and water. Insoluble in ether. 

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-35-5
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00149476
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
Synonym 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
PubChem CID 18530299
SMILES OC(=O)C1=NC2=CC=CC=C2C(O)=C1

Specifications

Melting Point ∼275°C (decomposition)
Quantity 5 g
Beilstein 147451
Merck Index 14,5327
Solubility Information Soluble in hot ethanol and water. Insoluble in ether.
IUPAC Name 4-hydroxyquinoline-2-carboxylic acid
Formula Weight 189.17 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.