missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinazoline, TRC

Product Code. 30400813
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400813 5 g 5g
30397716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400813 Supplier Toronto Research Chemicals Supplier No. TRCH9414155G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyquinazoline
CAS 491-36-1
Molecular Formula C8 H6 N2 O
Quantity 5 g
Synonym 4(1H)-Quinazolinone (6CI,7CI,8CI,9CI), 3,4-Dihydroquinazolin-4-one, 4(3H)-Quinazolone, 4-Hydroxyquinazoline, 4-Oxo-3,4-dihydroquinazoline, 4-Oxoquinazoline, 4-Quinazolinol, 4-Quinazolinone, 4-Quinazolone, C 201-0583, NSC 5863
InChI Formula InChI=1S/C8H6N2O/c11-8-6-3-1-2-4-7(6)9-5-10-8/h1-5H,(H,9,10,11)
SMILES O=C1N=CNC=2C=CC=CC12
IUPAC Name quinazolin-4-ol
Molecular Weight (g/mol) 146.15
Formula Weight 146.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.