missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypyridine, 95%

Product Code. 10071881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071881 100 g Glass bottle 100g
10062061 25 g Glass bottle 25g
10702905 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071881 Supplier Thermo Scientific Acros Supplier No. 348831000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-64-2 | C5H5NO | 95.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-64-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006419,MFCD00040458
InChI Key GCNTZFIIOFTKIY-UHFFFAOYSA-N
Synonym 4-hydroxypyridine, pyridin-4-ol, 4-pyridinol, 4-pyridone, 4 1h-pyridinone, pyridin-4 1h-one, 4-oxopyridine, 4-pyridinone, 4-pyridol, gamma-hydroxypyridine
PubChem CID 12290
IUPAC Name 1H-pyridin-4-one
SMILES O=C1C=CNC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 140.0°C to 147.0°C
Color Beige to Brown
Density 221g/mL
Boiling Point 230.0°C to 235.0°C (12.0 mmHg)
Flash Point 221°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula (C5H4N)OH
Quantity 100 g
Beilstein 21, V,7, 152
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol, slightly soluble in chloroform, pract. insoluble in ether and benzene
Formula Weight 95.1
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Hydroxypyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxypyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.