missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypyridine-2-carboxylic acid, 97%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
slightly soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
slightly soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽280°C (decomposition) |
| Quantity | 250 mg |
| Solubility Information | slightly soluble in water. |
| Formula Weight | 139.11 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Hydroxypyridine-2-carboxylic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction