missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxypropiophenone, 98+%, ACROS Organics™

Product Code. 10295060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
500g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10295060 500g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10295060 Supplier Thermo Scientific Acros Supplier No. 121815000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 70-70-2
CAS Min % 98.0
CAS Max % 100.0
Melting Point 147°C to 151°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Molecular Formula C9H10O2
Linear Formula HOC6H4COC2H5
MDL Number MFCD00002361
Quantity 500 g
Beilstein 08, 102
Merck Index 15, 7149
Synonym 4'-hydroxypropiophenone, paroxypropione, 4-hydroxypropiophenone, 1-4-hydroxyphenyl propan-1-one, mepal, paroxypropion, frenantol, frenohypon, p-hydroxypropiophenone, proxiphenon
InChI Key RARSHUDCJQSEFJ-UHFFFAOYSA-N
SMILES CCC(=O)C1=CC=C(C=C1)O
IUPAC Name 1-(4-hydroxyphenyl)propan-1-one
Molecular Weight (g/mol) 150.177
PubChem CID 6271
ChEBI CHEBI:32352
Formula Weight 150.18
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material 4';-Hydroxypropiophenone, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.