missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypiperidine, 99+%

Product Code. 10619092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10619092 100 g 100g
10130392 25 g 25g
11906331 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10619092 Supplier Thermo Scientific Acros Supplier No. 366511000
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 5382-16-1 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5382-16-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005999
InChI Key HDOWRFHMPULYOA-UHFFFAOYSA-N
Synonym 4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
PubChem CID 79341
IUPAC Name piperidin-4-ol
SMILES C1CNCCC1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 85.0°C to 90.0°C
Boiling Point 108.0°C to 114.0°C (10.0 mmHg)
Flash Point 107°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 21,I,188
Solubility Information Solubility in water: soluble.
Formula Weight 101.15
Percent Purity 99+%
Chemical Name or Material 4-Hydroxypiperidine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Hydroxypiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.