missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxypiperidine, 97%

Product Code. 11465377
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465377 100 g 100g
11445377 5 g 5g
11455377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465377 Supplier Thermo Scientific Alfa Aesar Supplier No. B22723.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5382-16-1 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5382-16-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00005999
InChI Key HDOWRFHMPULYOA-UHFFFAOYSA-N
Synonym 4-hydroxypiperidine, 4-piperidinol, 4-hydroxy piperidine, 4-hydroxypiperidin, piperidine-4-ol, piperdine-4-ol, piperid-4-ol, 4-hydoxypiperidine, 4-hydroxypiperdine, 4-hyroxypiperidine
PubChem CID 79341
IUPAC Name piperidin-4-ol
SMILES C1CNCCC1O

Specifications

Melting Point 86°C to 90°C
pH 11.5 to 11.7
Boiling Point 108°C to 114°C (10 mmHg)
Flash Point 107°C (224°F)
Odor Amine-like
Quantity 100 g
UN Number UN3263
Beilstein 102738
Sensitivity Hygroscopic
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 4-Hydroxypiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.