missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypiperidine-2,2,6,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxypiperidine-2,2,6,6-D4 |
| CAS | 1014695-51-2 |
| Molecular Formula | C5 D4 H7 N O |
| Quantity | 0.05 g |
| Synonym | 4-Piperidin-2,2,6,6-d4-ol (ACI), 2,2,6,6-Tetradeuteriopiperidin-4-ol, 4-Hydroxypiperidine-2,2,6,6-d4, 4-Hydroxypiperidine-2,2,6,6-D4, 4-Piperidinol-2,2,6,6-d4 |
| InChI Formula | InChI=1 S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7 H,1-4H2/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])CC(O)CC([2 H])([2 H])N1 |
| IUPAC Name | 2,2,6,6-tetradeuteriopiperidin-4-ol |
| Molecular Weight (g/mol) | 105.1716 |
| Formula Weight | 105.1092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction