missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetic acid, MedChemExpress

Product Code. 16479166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479166 Supplier Medchem Express Supplier No. HYN1902
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 4-hydroxyphenylacetic acid, a major microbiota-derived metabolite of polyphenols, is involved in the antioxidative action. 4-hydroxyphenylacetic acid induces expression of Nrf2.

4-hydroxyphenylacetic acid, a major microbiota-derived metabolite of polyphenols, is involved in the antioxidative action. 4-hydroxyphenylacetic acid induces expression of Nrf2.

Specifications

Chemical Name or Material 4-Hydroxyphenylacetic acid
CAS 156-38-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (657.25 mM)
SMILES O=C(O)CC1=CC=C(O)C=C1
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.