missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetaldehyde, approximately ~15% wt/vol in Ethyl Acetate, TRC

Product Code. 30390810
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390810 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390810 Supplier Toronto Research Chemicals Supplier No. TRCH94905050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyphenylacetaldehyde
CAS 7339-87-9
Purity Grade Notes GC
Molecular Formula C8 H8 O2
Quantity 50 mg
Synonym 4-Hydroxybenzeneacetaldehyde, (p-Hydroxyphenyl)acetaldehyde
InChI Formula InChI=1S/C8H8O2/c9-6-5-7-1-3-8(10)4-2-7/h1-4,6,10H,5H2
SMILES Oc1ccc(CC=O)cc1
IUPAC Name 2-(4-hydroxyphenyl)acetaldehyde
Molecular Weight (g/mol) 136.15
Formula Weight 136.0524
Percent Purity >95
Concentration or Composition (by Analyte or Components) 15 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.