missing translation for 'onlineSavingsMsg'
Learn More

(4-Hydroxyphenyl)-2-propionic acid, 98%, Thermo Scientific™

Product Code. 10155510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155510 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155510 Supplier Thermo Scientific Acros Supplier No. 302680050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 938-96-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 131.6°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H10O3
MDL Number MFCD00128847
Quantity 5 g
Synonym 2-4-hydroxyphenyl propanoic acid, 2-4-hydroxyphenyl propionic acid, 4-hydroxyhydratropate, 4-hydroxyhydratropic acid, 4-hydroxyphenyl-2-propionic acid, 2-4'-hydroxyphenyl-propionic acid, 2-4-hydroxyphenyl propionate, acmc-209ro0, 4-hydroxyphenyl propanoic acid, methyl 4-hydroxyphenylacetic acid
InChI Key ZHMMPVANGNPCBW-UHFFFAOYSA-N
SMILES CC(C1=CC=C(C=C1)O)C(=O)O
IUPAC Name 2-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 166.18
PubChem CID 102526
ChEBI CHEBI:1868
Formula Weight 166.18
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (4-Hydroxyphenyl)-2-propionic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.