missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxynonenal Antibody (198960) [DyLight 488], Novus Biologicals™

Product Code. 30731620 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731620 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731620 Supplier Novus Biologicals Supplier No. FAB3249K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

4-Hydroxynonenal Monoclonal antibody specifically detects 4-Hydroxynonenal in Broad Species Reactivity samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Hydroxynonenal
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 198960
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias (2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4-hydroxy-trans-2-nonenal
Host Species Mouse
Immunogen KLH-coupled 4-Hydroxynonenal
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.