missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxymidazolam, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
4-Hydroxymidazolam
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy Midazolam |
| CAS | 59468-85-8 |
| Analyte or Component Names | 4-Hydroxy Midazolam |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H13 Cl F N3 O |
| Synonym | 4 H-Imidazo[1,5-a][1,4 ]benzodiazepin-4-ol, 8-chloro-6-(2-fluorophenyl)-1-methyl- (9 CI, ACI), 8-Chloro-6-(2-fluorophenyl)-1-methyl-4 H-imidazo[1,5-a][1,4 ]benzodiazepin-4-ol (ACI), 4-Hydroxymidazolam |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cc1ncc2C(O)N=C(c3ccccc3F)c4cc(Cl)ccc4n12 |
| IUPAC Name | 8-chloro-6-(2-fluorophenyl)-1-methyl-4 H-imidazo[1,5-a][1,4 ]benzodiazepin-4-ol |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction