missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)tetrahydropyran, 98%

Product Code. 15441849
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441849 5 g 5g
15431849 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441849 Supplier Thermo Scientific Alfa Aesar Supplier No. H64568.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14774-37-9 | C6H12O2 | 116.16 g/mol

Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
SMILES OCC1CCOCC1

Specifications

Boiling Point 111°C to 115°C (15 mmHg)
Flash Point 61°C (142°F)
Refractive Index 1.46
Quantity 5 g
Beilstein 103069
Solubility Information Slightly soluble in water.
IUPAC Name (oxan-4-yl)methanol
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)tetrahydropyran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.