missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylacetic acid, 97%

Product Code. 11441810
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441810 5 g 5g
11431810 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441810 Supplier Thermo Scientific Alfa Aesar Supplier No. H27531.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73401-74-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73401-74-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065692
InChI Key FWZBPBKAANKOJQ-UHFFFAOYSA-N
Synonym 4-hydroxymethyl phenylacetic acid, 2-4-hydroxymethyl phenyl acetic acid, 4-hydroxymethyl phenyl acetic acid, 4-hydroxymethylphenylacetic acid, benzeneacetic acid,4-hydroxymethyl, benzeneacetic acid, 4-hydroxymethyl, ambotzrl-1037, pubchem19630, acmc-1bl5i
PubChem CID 3283498
IUPAC Name 2-[4-(hydroxymethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CO

Specifications

Melting Point 131°C to 134°C
Quantity 5 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.