missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenoxyacetic acid, 97%, Thermo Scientific™

Product Code. 11985131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11985131 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985131 Supplier Thermo Scientific Acros Supplier No. 441900050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 68858-21-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 107°C to 112°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C9H10O4
Quantity 5 g
Synonym 4-hydroxymethyl phenoxyacetic acid, 4-hydroxymethylphenoxyacetic acid, 2-4-hydroxymethyl phenoxy acetic acid, hmp linker, p-hydroxymethyl phenoxyacetic acid, 4-hydroxymethyl-phenoxy acetic acid
InChI Key VUCNQOPCYRJCGQ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CO)OCC(=O)O
IUPAC Name 2-[4-(hydroxymethyl)phenoxy]acetic acid
Molecular Weight (g/mol) 182.18
PubChem CID 194291
Formula Weight 182.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymethyl)phenoxyacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Hydroxymethyl)phenoxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.