missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl Pendimethalin, TRC

Product Code. 30391658
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391658 25 mg 25mg
30405318 250 mg -
30393727 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391658 Supplier Toronto Research Chemicals Supplier No. TRCH59406025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56750-76-6
Molecular Formula C13 H19 N3 O5
Molecular Weight (g/mol) 297.31
Synonym 4-[(1-Ethylpropyl)amino]-2-methyl-3,5-dinitrobenzenemethanol, 4-(Hydroxymethyl)pendimethalin, 4-[(1-Ethylpropyl)amino]-2-methyl-3,5-dinitrobenzyl alcohol, α4-Hydroxypendimethalin
IUPAC Name [2-methyl-3,5-dinitro-4-(pentan-3-ylamino)phenyl]methanol
SMILES CCC(CC)Nc1c(cc(CO)c(C)c1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H19N3O5/c1-4-10(5-2)14-12-11(15(18)19)6-9(7-17)8(3)13(12)16(20)21/h6,10,14,17H,4-5,7H2,1-3H3
Formula Weight 297.1325
Chemical Name or Material Pendimethalin-4-hydroxymethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.