missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl Pendimethalin, TRC

Product Code. 30405318
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405318 250 mg -
30391658 25 mg 25mg
30393727 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405318 Supplier Toronto Research Chemicals Supplier No. TRCH594060250MG
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56750-76-6
Molecular Formula C13 H19 N3 O5
Molecular Weight (g/mol) 297.31
Synonym 4-[(1-Ethylpropyl)amino]-2-methyl-3,5-dinitrobenzenemethanol, 4-(Hydroxymethyl)pendimethalin, 4-[(1-Ethylpropyl)amino]-2-methyl-3,5-dinitrobenzyl alcohol, α4-Hydroxypendimethalin
IUPAC Name [2-methyl-3,5-dinitro-4-(pentan-3-ylamino)phenyl]methanol
SMILES CCC(CC)Nc1c(cc(CO)c(C)c1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H19N3O5/c1-4-10(5-2)14-12-11(15(18)19)6-9(7-17)8(3)13(12)16(20)21/h6,10,14,17H,4-5,7H2,1-3H3
Formula Weight 297.1325
Chemical Name or Material Pendimethalin-4-hydroxymethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.