missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzonitrile, 97%

Product Code. 15274378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15274378 1 g 1g
15284378 5 g 5g
15294378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15274378 Supplier Thermo Scientific Acros Supplier No. 454650010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 874-89-5 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00870633
InChI Key XAASLEJRGFPHEV-UHFFFAOYSA-N
Synonym 4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile
PubChem CID 160549
IUPAC Name 4-(hydroxymethyl)benzonitrile
SMILES OCC1=CC=C(C=C1)C#N

Specifications

Melting Point 41.0°C to 42.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 4-(Hydroxymethyl)benzonitrile
compliance-icons
Product Identifier
  • 4-(Hydroxymethyl)benzonitrile, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.