missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)benzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11419727
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3006-96-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00017598
InChI Key WWYFPDXEIFBNKE-UHFFFAOYSA-N
Synonym 4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd
PubChem CID 76360
IUPAC Name 4-(hydroxymethyl)benzoic acid
SMILES OCC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 179°C to 183°C
Odor Odorless
Quantity 25 g
Beilstein 2690023
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 4-(Hydroxymethyl)benzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'