missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-oxide, 98%, Thermo Scientific Chemicals

Product Code. 15496846
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5301-78-0
Molecular Formula C5H9O5P
Molecular Weight (g/mol) 180.096
MDL Number MFCD01677612
InChI Key YASRHLDAFCMIPB-UHFFFAOYSA-N
PubChem CID 199942
IUPAC Name (1-oxo-2,6,7-trioxa-1$l^{5}-phosphabicyclo[2.2.2]octan-4-yl)methanol
SMILES C1C2(COP(=O)(O1)OC2)CO

Specifications

Color White
Quantity 5 g
Synonym 4-hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo 2.2.2 octane 1-oxide, pentaerythritol phosphate alcohol, 2,6,7-trioxa-1-phosphabicyclo2.2.2octane-4-methanol, 1-oxide, pentaerythritol, cyclic phosphate 1:1 8ci, 2,6,7-trioxa-1-phosphabicyclo-2.2.2.-octane-4-methanol 1-oxide, 4-hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo 2.2.2 octane-1-oxide, 2,6,7-trioxa-1-phosphabicyclo 2.2.2 octane-4-methanol 1-oxide, 2,6,7-trioxa-1-phosphabicyclo 2.2.2 octane-4-methanol, 1-oxide, 4-hydroxymethyl-2,6,7-trioxa-1??-phosphabicyclo 2.2.2 octan-1-one
Formula Weight 180.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Hydroxymethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'