missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymandelic Acid Monohydrate, TRC

Product Code. 30400039
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400039 100 g
30391352 2.5 g
30401057 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400039 Supplier Toronto Research Chemicals Supplier No. TRCH944355100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184901-84-6
Molecular Formula C8H8O4 . H2O
Molecular Weight (g/mol) 186.16
Synonym alpha,4-Dihydroxybenzeneacetic Acid
IUPAC Name 2-hydroxy-2-(4-hydroxyphenyl)acetic acid;hydrate
SMILES O.OC(C(=O)O)c1ccc(O)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H8O4.H2O/c9-6-3-1-5(2-4-6)7(10)8(11)12;/h1-4,7,9-10H,(H,11,12);1H2
Formula Weight 186.0528
Chemical Name or Material 4-Hydroxymandelic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.