missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyisovaleric Acid Sodium Salt, TRC

Product Code. 30395724
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395724 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395724 Supplier Toronto Research Chemicals Supplier No. TRCH812890100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyisovaleric Acid Sodium Salt
CAS 112895-77-9
Molecular Formula C5H9NaO3
Quantity 100 mg
Synonym 4-Hydroxy-3-methyl-butanoic Acid Monosodium Salt
InChI Formula InChI=1S/C5H10O3.Na/c1-4(3-6)2-5(7)8;/h4,6H,2-3H2,1H3,(H,7,8);/q;+1/p-1
SMILES [Na].O=C(O)CC(C)CO
Molecular Weight (g/mol) 140.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.