missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxyisophthalic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxyisophthalic Acid |
| CAS | 636-46-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H6 O5 |
| Quantity | 250 mg |
| Synonym | 4-Hydroxybenzene-1,3-dicarboxylic Acid, 4-Hydroxyisophthalic Acid, Acetylsalicylic Acid Imp. B (EP), Salicylic Acid Imp. B (EP), 1,3-Benzenedicarboxylic acid, 4-hydroxy-, Isophthalic acid, 4-hydroxy- (6CI,7CI,8CI), 4-Hydroxy-1,3-benzenedicarboxylic acid, 4-Hydroxy-m-phthalic acid, Eupirina, NSC 2445 |
| InChI Formula | InChI=1S/C8H6O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H,(H,10,11)(H,12,13) |
| SMILES | OC(=O)c1ccc(O)c(c1)C(=O)O |
| IUPAC Name | 4-hydroxybenzene-1,3-dicarboxylic acid |
| Molecular Weight (g/mol) | 182.13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?