missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyindole O-b-D-Glucuronide Sodium Salt, TRC

Product Code. 30399734
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399734 25 mg 25mg
30391224 2.5 mg 2.5mg
30397921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399734 Supplier Toronto Research Chemicals Supplier No. TRCH94344325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxyindole O-Beta-D-Glucuronide Sodium Salt
Analyte or Component Names 4-Hydroxyindole O-Beta-D-Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C14 H14 N Na O7
Synonym Sodium (2S,3S,4S,5R,6S)-6-((1H-indol-4-yl)oxy)-3,4,5-Trihydroxytetrahydro-2H-pyran-2-carboxylate
InChI Formula InChI=1S/C14H15NO7.Na/c16-9-10(17)12(13(19)20)22-14(11(9)18)21-8-3-1-2-7-6(8)4-5-15-7;/h1-5,9-12,14-18H,(H,19,20);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1
Solution Type Neat
SMILES O[C@H]1[C@H](Oc2cccc3[nH]ccc23)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O[Na]
IUPAC Name [(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(1H-indol-4-yloxy)oxane-2-carbonyl]oxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 331.253
Quantity 25 mg
Formula Weight 331.067
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.