missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyindole, 98%

Product Code. 15499009
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15499009

Brand: Thermo Scientific Alfa Aesar L14352.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2380-94-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-94-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005667
InChI Key NLMQHXUGJIAKTH-UHFFFAOYSA-N
Synonym 4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
PubChem CID 75421
ChEBI CHEBI:24702
IUPAC Name 1H-indol-4-ol
SMILES OC1=C2C=CNC2=CC=C1

Specifications

Melting Point 97°C to 99°C
Quantity 25 g
Beilstein 114905
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxyindole
compliance-icons
Product Identifier
  • 4-Hydroxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-Hydroxyindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.