missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyflavanone, TRC

Product Code. 30405837
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30405837 25 mg -
30390858 250 mg 250mg
30395424 50 mg -
3 options
This item is not returnable. View return policy

Product Code. 30405837

Brand: Toronto Research Chemicals TRCH94244325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6515-37-3
Molecular Formula C15 H12 O3
Molecular Weight (g/mol) 240.25
Synonym (±)-4'-Hydroxyflavanone
IUPAC Name 2-(4-hydroxyphenyl)chroman-4-one
SMILES Oc1ccc(cc1)C2CC(=O)c3ccccc3O2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H12O3/c16-11-7-5-10(6-8-11)15-9-13(17)12-3-1-2-4-14(12)18-15/h1-8,15-16H,9H2
Formula Weight 240.0786
Chemical Name or Material 4'-Hydroxyflavanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.