missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycyclohexanone ethylene acetal, 90+%

Product Code. 15483347
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483347 Supplier Thermo Scientific Alfa Aesar Supplier No. H26037.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22428-87-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22428-87-1
Molecular Formula C8H14O3
Refractive Index 1.488
MDL Number MFCD00067003
Quantity 1 g
Synonym 1,4-dioxaspiro 4.5 decan-8-ol, 1,4-dioxa-spiro 4.5 decan-8-ol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, pubchem20110, acmc-1chv5, 4,4-ethylenedioxycyclohexanol
InChI Key HKQTYQDNCKMNHZ-UHFFFAOYSA-N
SMILES OC1CCC2(CC1)OCCO2
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-ol
Molecular Weight (g/mol) 158.20
PubChem CID 567352
Formula Weight 158.19
Percent Purity ≥90%
Chemical Name or Material 4-Hydroxycyclohexanone ethylene acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.