missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin, LoGiCal

Product Code. 30458582
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458582 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458582 Supplier LoGiCal Supplier No. LGCFOR0732.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxycoumarin
CAS 1076-38-6
Analyte or Component Names 4-Hydroxycoumarin
Shipping Condition Room Temperature
Molecular Formula C9 H6 O3
Certifications/Compliance ISO 9001
Synonym 4-Hydroxy-2H-1-benzopyran-2-one (4-Hydroxycoumarin), Warfarin Sodium Imp. B (EP), Warfarin Imp. B (EP), 4-Hydroxy-2H-1-benzopyran-2-one, 4-Hydroxycoumarin, 4-Coumarinol, 4-Hydroxy-2H-benzo[b]pyran-2-one, 4-Hydroxychromen-2-one, Benzotetronic acid, NSC 11889, Warfarin Sodium Impurity B, Warfarin Impurity B
InChI Formula InChI=1S/C9H6O3/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5,10H
Solution Type Neat
SMILES OC1=CC(=O)Oc2ccccc12
IUPAC Name 4-hydroxychromen-2-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 162.14
Quantity 10 mg
Formula Weight 162.0317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.