missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxychalcone, 97%, Thermo Scientific Chemicals

Catalog Number 11459802
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2657-25-2
C15H12O2
224.259
MFCD00016484
UAHGNXFYLAJDIN-IZZDOVSWSA-N
4'-hydroxychalcone, chalcone, 4'-hydroxy, 1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, 2e-1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, p-cinnamoylphenol, 2-benzal-4'-hydroxyacetophenone, hydroxychalcone, 4', 2-propen-1-one, 1-4-hydroxyphenyl-3-phenyl, unii-2k338k8uoa, e-1-4-hydroxyphenyl-3-phenyl-prop-2-en-1-one
5282362
CHEBI:34360
(E)-1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one
C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)O
This item is not returnable. View return policy
2657-25-2
C15H12O2
224.259
MFCD00016484
UAHGNXFYLAJDIN-IZZDOVSWSA-N
4'-hydroxychalcone, chalcone, 4'-hydroxy, 1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, 2e-1-4-hydroxyphenyl-3-phenylprop-2-en-1-one, p-cinnamoylphenol, 2-benzal-4'-hydroxyacetophenone, hydroxychalcone, 4', 2-propen-1-one, 1-4-hydroxyphenyl-3-phenyl, unii-2k338k8uoa, e-1-4-hydroxyphenyl-3-phenyl-prop-2-en-1-one
5282362
CHEBI:34360
(E)-1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one
C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175°C to 177°C
Quantity 5 g
Beilstein 2049150
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material 4'-Hydroxychalcone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'