missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycarbazeran, TRC

Product Code. 30004181
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004181 10 mg 10mg
30011310 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004181 Supplier Toronto Research Chemicals Supplier No. TRCH88412510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96724-43-5
Molecular Formula C18H24N4O5
Molecular Weight (g/mol) 376.41
Synonym 1-(3,4-Dihydro-6,7-dimethoxy-4-oxo-1-phthalazinyl)-4-piperidinyl Ester Ethylcarbamic Acid
IUPAC Name 1-(6,7-dimethoxy-4-oxo-3,4-dihydrophthalazin-1-yl)piperidin-4-yl ethylcarbamate
SMILES CCNC(=O)OC1CCN(CC1)C2=NNC(=O)C3=CC(=C(C=C32)OC)OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H24N4O5/c1-4-19-18(24)27-11-5-7-22(8-6-11)16-12-9-14(25-2)15(26-3)10-13(12)17(23)21-20-16/h9-11H,4-8H2,1-3H3,(H,19,24)(H,21,23)
Formula Weight 376.17
Percent Purity >95%
Chemical Name or Material 4-Hydroxycarbazeran
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.