missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybutyl acrylate, 95%, stab. with ca 500ppm 4-methoxyphenol

Product Code. 11377546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377546 5 g 5g
11387546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377546 Supplier Thermo Scientific Alfa Aesar Supplier No. L11097.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2478-10-6 | C7H16ClNO2S | 213.72 g/mol

4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2478-10-6
MDL Number MFCD00010261

Specifications

Melting Point -112°C
Density 1.041
Boiling Point ∼230°C
Flash Point >110°C (230°F)
Molecular Formula C7H12O3
Refractive Index 1.452
Quantity 5 g
UN Number UN3082
Beilstein 1756545
Synonym 4-hydroxybutyl acrylate, acrylic acid 4-hydroxybutyl ester, hydroxybutyl acrylate, 1,4-butanediol monoacrylate, 2-propenoic acid, 4-hydroxybutyl ester, unii-4o2a4het1x, 4o2a4het1x, hydroxybutylacrylate, 4-hydroxybutylacrylate, 4-hydroxy-butyl acrylate
Solubility Information Fully miscible in water.
InChI Key NDWUBGAGUCISDV-UHFFFAOYSA-N
SMILES C=CC(=O)OCCCCO
IUPAC Name 4-hydroxybutyl prop-2-enoate
Molecular Weight (g/mol) 144.17
PubChem CID 75588
Formula Weight 144.17
Percent Purity 95%
Chemical Name or Material 4-Hydroxybutyl acrylate, stabilized with ≈500ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.