missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybut-2-ynoic Acid, TRC

Product Code. 30401295
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401295 100 mg -
30402173 50 mg 50mg
30390388 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401295 Supplier Toronto Research Chemicals Supplier No. TRCH977653100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7218-52-2
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
IUPAC Name 4-hydroxybut-2-ynoic acid
SMILES OCC#CC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H4O3/c5-3-1-2-4(6)7/h5H,3H2,(H,6,7)
Formula Weight 100.016
Chemical Name or Material 4-Hydroxybut-2-ynoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.