missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzylidenemalononitrile, 98%, Thermo Scientific™

Product Code. 11371647
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371647 10 g 10g
11381647 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371647 Supplier Thermo Scientific Alfa Aesar Supplier No. L04651.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3785-90-8
Molecular Formula C10H6N2O
Molecular Weight (g/mol) 170.17
MDL Number MFCD00020189
InChI Key FNCOVSWSZZVFBQ-UHFFFAOYSA-N
Synonym tyrphostin 8, 4-hydroxybenzylidenemalononitrile, malononitrile, p-hydroxybenzylidene, 2-4-hydroxyphenyl methylidene propanedinitrile, 4-hydroxybenzylidene malononitrile, 4-hydroxy-benzal-malonitril german, propanedinitrile, 4-hydroxyphenyl methylene, 4-hydroxybenzylidene propanedinitrile, 2-4-hydroxybenzylidene malononitrile
PubChem CID 92970
IUPAC Name 2-[(4-hydroxyphenyl)methylidene]propanedinitrile
SMILES OC1=CC=C(C=C(C#N)C#N)C=C1

Specifications

Melting Point 187°C to 190°C
Quantity 10 g
UN Number UN3439
Beilstein 2209079
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzylidenemalononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.