missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl Methanethiosulfonate, TRC

Product Code. 30407843
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407843 2.5 mg 2.5mg
30393388 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407843 Supplier Toronto Research Chemicals Supplier No. TRCH8297502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzyl Methanethiosulfonate
CAS 491868-12-3
Molecular Formula C8 H10 O3 S2
Quantity 2.5 mg
InChI Formula InChI=1S/C8H10O3S2/c1-13(10,11)12-6-7-2-4-8(9)5-3-7/h2-5,9H,6H2,1H3
SMILES CS(=O)(=O)SCc1ccc(O)cc1
IUPAC Name 4-(methylsulfonylsulfanylmethyl)phenol
Molecular Weight (g/mol) 218.29
Formula Weight 218.0071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.