missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl Alcohol, Mikromol™

Product Code. 30463738
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463738 4 x 25 mg 100mg
17993546 25 mg 25mg
17951776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463738 Supplier Mikromol Supplier No. MM0460.0600254AMP
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Hydroxybenzyl Alcohol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzyl Alcohol
CAS 623-05-2
Analyte or Component Names 4-Hydroxybenzyl Alcohol
Shipping Condition Room Temperature
Molecular Formula C7 H8 O2
Certifications/Compliance ISO 17025
Synonym 4-Hydroxybenzyl Alcohol
InChI Formula InChI=1S/C7H8O2/c8-5-6-1-3-7(9)4-2-6/h1-4,8-9H,5H2
Solution Type Neat
SMILES OCc1ccc(O)cc1
IUPAC Name 4-(hydroxymethyl)phenol
Molecular Weight (g/mol) 124.14
Quantity 4 x 25 mg
Formula Weight 124.0524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.